missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Fructose, 99%

D(-)-Fructose, 99%, C6H12O6, CAS Number-57-48-7, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, arabino-hexulose, d--fructose, d---fructose, d-levulose, fructose, d, furucton, keto-d-fructose, krystar 300, sugar, fruit, 500g, Passes test, 98.5% min, 31,321, CHEBI:48095, 50ppm max, White, 200-333-3

49.35 € - 492.45 €

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10307440
View Documents
Thermo Scientific Chemicals
161355000
500 g Plastic bottle
49.35 €
500g
Please to purchase this item. Need a web account? Register with us today!
10397430
View Documents
Thermo Scientific Chemicals
161350010
1 kg Plastic bottle
83.82 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10634642
View Documents
Thermo Scientific Chemicals
161350025
2.5 kg Plastic bottle
164.82 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
11984041
View Documents
Thermo Scientific Chemicals
161350100
10 kg Plastic drum
492.45 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

57-48-7
180.16
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one

Specifications

57-48-7
White
Authentic
C6H12O6
15,4302
− 92.25
Solubility in water: 3750g/L (20°C). Other solubilities: soluble in alcohol,pyridine,ether,ethylamine,and methylamine
C(C(C(C(C(=O)CO)O)O)O)O
180.16
CHEBI:48095
99%
D(-)-Fructose
103.0°C to 105.0°C
0.5% (70°C, 4 hrs) (vacuum) max.
98.5% min
31,321
− 92.25 (20.00°C c=10,H2O,on dry sub.)
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-333-3

Product Identifier
  • D(-)-Fructose

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only