missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Fructose, Pure, Fisher Chemical™

Product Code. 10346040
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D(-)-Fructose, Pure

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number 148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 103°C
Color White
pH 5 to 7
Packaging Plastic powder jar
Quantity 250 g
Formula Weight 180.16g/mol
Physical Form Solid
Product Identifier
  • D(-)-Fructose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.