missing translation for 'onlineSavingsMsg'
Learn More

D-Galacto-D-mannan, Thermo Scientific Chemicals

Product Code. 15408039
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11078-30-1
MDL Number MFCD00146683
Synonym galactomannan, galactomannoglycan, d-galacto-d-mannane, 6-o-alpha-d-galactopyranosyl-4-o-beta-d-mannopyranosyl-beta-d-mannopyranose, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxyoxan-2-yl methoxy oxane-3,4,5-triol, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-3-2s,3s,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-tetrahydropyran-2-yl methoxy tetrahydropyran-3,4,5-triol, 2r,3s,4s,5s,6s-2-hydroxymethyl-6-2r,3s,4r,5s,6r-4,5,6-trihydroxy-2-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy oxane-3,4,5-triol

Specifications

Color White
Quantity 100 mg
Formula Weight ∽200000
Physical Form Powder
Chemical Name or Material D-Galacto-D-mannan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.