missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Galactose, 99+%, Thermo Scientific Chemicals

D-Galactose, CAS # 59-23-4, is a right-handed conformation of a monosaccharide that is grouped as an aldohexose, and the molecule may be useful in study of aging, inflammation, and metabolic disorders.

62.21 € - 4961.69 €

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00063833
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10011560
View Documents
Thermo Scientific Acros
150611000
100 g
62.21 €
100g
Please to purchase this item. Need a web account? Register with us today!
10307470
View Documents
Thermo Scientific Acros
150615000
500 g
216.08 €
500g
Please to purchase this item. Need a web account? Register with us today!
10427360
View Documents
Thermo Scientific Acros
150610010
1 kg
419.09 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10649283
View Documents
Thermo Scientific Acros
150610051
5 kg
1483.38 €
5kg
Please to purchase this item. Need a web account? Register with us today!
15224644
View Documents
Thermo Scientific Acros
150610250
25 kg
4961.69 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Galactose is a monosaccharide and aldohexose found in most organisms
  • Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
  • It also plays a role as a metabolite in mice and E. coli
  • Galactose is less stable than glucose and rapid conversion often occurs
  • Galactose may also be incorporated into glycoconjugates

Applications

  • Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
  • It is intended for a variety of applications from in vitro metabolic assays to HPLC
Specifications

Chemical Identifiers

59-23-4
C6H12O6
MFCD00063833

Specifications

59-23-4
White
Authentic
C6H12O6
31, 295
+ 79.75 (20.00°C c=10,H2O,NH3)
180.16
≥99%
D(+)-Galactose
165.0°C to 168.0°C
6.2
99+%
MFCD00063833
15, 4365
+ 79.75
Negligible
Powder
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.