missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose, ACS Reagent, anhydrous

Product Code. 10391201 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10391201

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50-99-7 | C6H12O6 | 180.156 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
MDL Number MFCD00063684
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.0°C
Color White
Ignition Residue 0.02% max.
Loss on Drying 0.2% max. (105°C, 6 hrs)
Infrared Spectrum Authentic
Packaging Plastic Bottle
Molecular Formula C6H12O6
Quantity 500 g
Beilstein 31, 83
Merck Index 15, 4495
Specific Rotation Condition + 52.75 (25.00°C c=10,H2O,NH4OH)
Specific Rotation + 52.75
Solubility Information 0.005% max. Insoluble Matter
Molecular Weight (g/mol) 180.156
Formula Weight 180.16
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Glucose, Anhydrous, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.