missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose, anhydrous, 99%

Product Code. 11472858
Click to view available options
Quantity:
10,000 g
2500 g
500 g

D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucose is used in various metabolic processes including enzymic synthesis of cyclohexyl-alpha and beta -D-glucosides. It is involved in the detection of type 2 diabetes mellitus and the most widely used aldohexose in living organisms. Its availability influences psychological processes since it is a primary energy source for the brain.

Solubility
Soluble in water, hot glacial acetic acid, pyridine, aniline and dimethyl sulfoxide. Slightly soluble in methanol, ethanol and acetone.

Notes
Store in cool place. Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00063774
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 147°C to 153°C
Assay Percent Range 99%
Quantity 2500 g
Beilstein 1281608
Merck Index 14,4459
Solubility Information Soluble in water,hot glacial acetic acid,pyridine,aniline and dimethyl sulfoxide. Slightly soluble in methanol,ethanol and acetone.
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-(+)-Glucose, anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.