missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | 146.0°C |
| Loss on Drying | 0.5% max. (1 g, 105°C, 16 h) |
| Quantity | 5 kg |
| Merck Index | 15,4495 |
| Specific Rotation Condition | + 52.75 (25.00°C c=10, H2O, NH4OH) |
| Specific Rotation | + 52.75 |
| Solubility Information | Solubility in water: soluble. |
| Formula Weight | 180.16 |
| Percent Purity | 97.5 to 102.0% (Anhydrous substance) |
| Related Substances | 0.5% max. (total impurities) |
| Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur