missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-D-Glucose pentaacetate, 98%

Product Code. 10096193 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10096193

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 604-69-3 | C16H22O11 | 390.341 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 604-69-3
MDL Number MFCD00006597
InChI Key LPTITAGPBXDDGR-IBEHDNSVSA-N
Synonym beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β
PubChem CID 2724702
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C16H22O11
Quantity 500 g
Specific Rotation Condition + 5.00 (20.00°C c=1,CHCL3)
Specific Rotation + 5.00
Molecular Weight (g/mol) 390.341
Formula Weight 390.33
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material β-D-Glucose pentaacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.