missing translation for 'onlineSavingsMsg'
Learn More
D-Glucuronic acid, 98%, Thermo Scientific Chemicals Product Code.: 10472071

D-Glucuronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10472071
25 g, Plastic bottle
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10472071

Brand: Thermo Scientific Acros 309880250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6556-12-3
Molecular Formula C6H10O7
Molecular Weight (g/mol) 194.14
MDL Number MFCD00064368
InChI Key IAJILQKETJEXLJ-QTBDOELSSA-N
ChEBI CHEBI:47953
IUPAC Name (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

Specifications

Melting Point 158.0°C to 162.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Merck Index 15,4501
Specific Rotation Condition + 36.50 (20.00°C c=6,H2O,after 2 hrs)
Specific Rotation + 36.50
Synonym 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoic acid, aldehydo-d-glucuronic acid, unii-8a5d83q4rw, rel, d-glucuronic acid, d-+ glucuronic acid, d03jeu, d-glucuronic acid 5g
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O
PubChem CID 65041
Formula Weight 194.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Glucuronic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Glucuronic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.