missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucuronic acid, sodium salt, monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10627304 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10627304

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14984-34-0 | C6H9NaO7·H2O | 234.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14984-34-0
Molecular Formula C6H9NaO7·H2O
Molecular Weight (g/mol) 234.14
MDL Number MFCD00151051
InChI Key QKHMTHNLNZGTSP-JSCKKFHOSA-N
Synonym sodium d-glucuronate
PubChem CID 87235405
IUPAC Name sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na]

Specifications

Melting Point 134°C to 140°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 100 g
Beilstein 03, 886
Merck Index 10, 4327
Specific Rotation Condition +21° (20°C c=2,H2O,after 4hrs.)
Specific Rotation + 21
Formula Weight 234.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Glucuronic acid, sodium salt, monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.