missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Lactose, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15547994
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 64044-51-5
Packaging Poly Bottle
Molecular Formula C12H24O12
Quantity 500 g
Specific Gravity 1L = 1.53kg
Synonym d-lactose monohydrate, d-+-lactosemonohydrate
InChI Key WSVLPVUVIUVCRA-ZEAJSSQNSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
IUPAC Name (3S,6R)-2-(hydroxymethyl)-6-[(3R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Molecular Weight (g/mol) 360.312
PubChem CID 133126686
Formula Weight 360.32
Grade BAKER ANALYZED™ ACS Reagent
Physical Form Powder
Chemical Name or Material D-(+)-Lactose, Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'