missing translation for 'onlineSavingsMsg'
Learn More
D-Luciferin sodium salt monohydrate, 98+%, Thermo Scientific Chemicals Product Code.: 10668254

D-Luciferin sodium salt monohydrate, 98+%, Thermo Scientific Chemicals

Product Code. 10668254
10 mg, Glass bottle
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10668254

Brand: Thermo Scientific Acros 336780100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103404-75-7
Molecular Formula C11H7N2NaO3S2
Molecular Weight (g/mol) 302.298
MDL Number MFCD11865368
InChI Key BZNVUYVALNTPBG-QWLWRJJTSA-M
Synonym Firefly Luciferin monosodium salt monohydrate
PubChem CID 133109097
IUPAC Name sodium;(2E,4R)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate
SMILES C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)[O-].[Na+]

Specifications

Assay Percent Range 98+%
Infrared Spectrum Authentic
Formula Weight 320.32
Percent Purity ≥98.0%
Packaging Glass bottle
Specific Extinction 17500 min. (385nm, 0.5M Na2CO3) min. (pH 11.5)
Color Yellow
Quantity 10 mg
Physical Form Powder
Chemical Name or Material D-Luciferin sodium salt monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0331613 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number336780500,336780100
Product Identifier
  • D-Luciferin sodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-Luciferin sodium salt monohydrate, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.