missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Luciferin sodium salt monohydrate, 98+%, Thermo Scientific Chemicals
213.00 € - 761.00 €
Chemical Identifiers
CAS | 103404-75-7 |
---|---|
Molecular Formula | C11H7N2NaO3S2 |
Molecular Weight (g/mol) | 302.298 |
MDL Number | MFCD11865368 |
InChI Key | BZNVUYVALNTPBG-QWLWRJJTSA-M |
Synonym | Firefly Luciferin monosodium salt monohydrate |
PubChem CID | 133109097 |
IUPAC Name | sodium;(2E,4R)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate |
SMILES | C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10668254
|
Thermo Scientific Acros
336780100 |
10 mg | Glass bottle |
213.00 €
10mg |
In Stock
Log in to see stock. |
||||
10758942
|
Thermo Scientific Acros
336780500 |
50 mg | Glass bottle |
761.00 €
50mg |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103404-75-7 | |
302.298 | |
BZNVUYVALNTPBG-QWLWRJJTSA-M | |
133109097 | |
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)[O-].[Na+] |
C11H7N2NaO3S2 | |
MFCD11865368 | |
Firefly Luciferin monosodium salt monohydrate | |
sodium;(2E,4R)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate |
Specifications
103404-75-7 | |
C11H7N2NaO3S2 | |
Firefly Luciferin monosodium salt monohydrate | |
Authentic | |
sodium;(2E,4R)-2-(6-oxo-1,3-benzothiazol-2-ylidene)-1,3-thiazolidine-4-carboxylate | |
133109097 | |
≥98.0% | |
Yellow | |
D-Luciferin sodium salt monohydrate |
98+% | |
MFCD11865368 | |
BZNVUYVALNTPBG-QWLWRJJTSA-M | |
C1C(NC(=C2N=C3C=CC(=O)C=C3S2)S1)C(=O)[O-].[Na+] | |
302.298 | |
320.32 | |
17500 min. (385nm, 0.5M Na2CO3) min. (pH 11.5) | |
Powder |
Safety and Handling
AIR SENSITIVE
MOISTURE SENSITIVE
Product Identifier
- D-Luciferin sodium salt monohydrate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title