missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Malic acid, 98%

Product Code. 10503131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10503131

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 636-61-3 | C4H6O5 | 134.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-61-3
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.09
MDL Number MFCD00004245
InChI Key BJEPYKJPYRNKOW-UHFFFAOYNA-N
Synonym d-+-malic acid, d-malic acid, d +-malic acid, r-2-hydroxysuccinic acid, r-malic acid, malic acid, d, 2r-2-hydroxybutanedioic acid, l +-malic acid, d-malate
PubChem CID 92824
ChEBI CHEBI:30796
IUPAC Name (2R)-2-hydroxybutanedioic acid
SMILES OC(CC(O)=O)C(O)=O

Specifications

Melting Point 99.0°C to 104.0°C
Color White
Assay Percent Range 98%
Linear Formula HO2CCH2CH(OH)CO2H
Beilstein 03, 417
Merck Index 15, 5771
Solubility Information Solubility in water: soluble.
Formula Weight 134.09
Percent Purity 98%
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Malic acid
Show More Show Less
compliance-icons
Product Identifier
  • D(+)-Malic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.