missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannitol, 98+%

Product Code. 10488433
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 165.0°C to 170.0°C
Color White
Loss on Drying 0.3% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 2.5 kg
Beilstein 01, IV, 2841
Merck Index 15, 5810
Specific Rotation +23° to +25° (20°C, 589nm) (PHEUR)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and aniline,slightly soluble in lower alcohols,practically insoluble in most other common,organic solvents
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Formula Weight 182.17
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material D-Mannitol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.