missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Melibiose monohydrate, 99+%

Product Code. 11976891 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11976891

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66009-10-7 | C12H24O12 | 360.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66009-10-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00198188
InChI Key CHIDEFLSUMQFBY-UHFFFAOYNA-N
Synonym d +-melibiose monohydrate, d-+-melibiose hydrate, 585-99-9 anhydrous, melibiose hplc, 6-, a-d-galactopyranosyl-d-glucopyranose, 6-o-a-d-galactopyranosyl-d-glucopyranose hydrate, 6-o-a-d-galactopyranosyl-d-glucose monohydrate, 6-o-a-d-galactopyranosyl-d-glucopyranose monohydrate, 6-o-alpha-d-galactopyranosyl-d-glucose monohydrate, d-glucose, 6-o-a-d-galactopyranosyl-, monohydrate 9ci
PubChem CID 71308738
IUPAC Name (2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyhexanal;hydrate
SMILES O.OCC1OC(OCC(O)C(O)C(O)C(O)C=O)C(O)C(O)C1O

Specifications

Acidity 0.4mL 0.1N NaOH/6g max.
Melting Point 182.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Odor Odorless
Packaging Glass bottle
Quantity 500 g
Beilstein 31,421
Merck Index 14,582
Specific Rotation Condition + 135.00 (20.00°C c=4,H2O/NH3)
Specific Rotation + 135.00
Solubility Information Solubility in water: soluble
Formula Weight 360.32
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Melibiose monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.