missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Methionine, 99%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.
Solubility
Soluble in water. 53 g/L (20°C).
Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.
Solubility
Soluble in water. 53 g/L (20°C).
Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
Specifications
Specifications
| Melting Point | ∼280°C (decomposition) |
| Beilstein | 1722293 |
| Merck Index | 14,5975 |
| Solubility Information | Soluble in water. 53g/L (20°C). |
| Quantity | 25 g |
| Formula Weight | 149.21 |
| Optical Rotation | −23° (c=2 in 2N HCl) |
| Percent Purity | 99% |
| Chemical Name or Material | D-Methionine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction