Learn More
D-Panthenol, 98+%, Thermo Scientific Chemicals
54.27 € - 178.89 €
Chemical Identifiers
CAS | 81-13-0 |
---|---|
Molecular Formula | C9H19NO4 |
Molecular Weight (g/mol) | 205.25 |
MDL Number | MFCD00065006 |
InChI Key | SNPLKNRPJHDVJA-ZETCQYMHSA-N |
Synonym | dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton |
PubChem CID | 131204 |
ChEBI | CHEBI:27373 |
IUPAC Name | (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide |
SMILES | CC(C)(CO)[C@@H](O)C(=O)NCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11418417
|
Thermo Scientific Alfa Aesar
A18499.22 |
100 g |
54.27 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428417
|
Thermo Scientific Alfa Aesar
A18499.36 |
500 g |
178.89 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.
Solubility
Fully miscible in water and soluble in alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Chemical Identifiers
81-13-0 | |
205.25 | |
SNPLKNRPJHDVJA-ZETCQYMHSA-N | |
131204 | |
(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide |
C9H19NO4 | |
MFCD00065006 | |
dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton | |
CHEBI:27373 | |
CC(C)(CO)[C@@H](O)C(=O)NCCCO |
Specifications
81-13-0 | |
118°C to 120°C (0.02 mmHg) | |
C9H19NO4 | |
MFCD00065006 | |
Hygroscopic | |
dexpanthenol, d-panthenol, pantothenol, bepanthen, d-pantothenyl alcohol, ilopan, +-panthenol, bepanthene, panthoderm, thenalton | |
SNPLKNRPJHDVJA-ZETCQYMHSA-N | |
CC(C)(CO)[C@@H](O)C(=O)NCCCO | |
205.25 | |
CHEBI:27373 | |
≥98% |
1.162 | |
159°C (318°F) | |
1.499 | |
1724947 | |
14,2947 | |
Fully miscible in water and soluble in alcohol. | |
−30° (c=5 in Water) | |
(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide | |
131204 | |
205.26 | |
D-Panthenol |
Safety and Handling
missing translation for 'einecsNumber' : 201-327-3
missing translation for 'rtecsNumber' : ES4316000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.