missing translation for 'onlineSavingsMsg'
Learn More

D-Phenylalaninol, 98%, Thermo Scientific Chemicals

Product Code. 11342217
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Phenylalaninol is used as a chiral auxiliary for asymmetric Michael reactions. It is an enantiomer of L-Phenylalaninol, an inhibitor of intestinal Phenylalanine absorption. It acts as an inhibiting agent to the enzymes which are responsible for the breakdown of endorphins.

Solubility
Soluble in Dichloromethane, Ethyl Acetate, Methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with carbondioxide, air, acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 5267-64-1
Molecular Formula C9H13NO
MDL Number MFCD00064298
Synonym d-phenylalaninol, r-2-amino-3-phenylpropan-1-ol, d-penylalaninol, r-+-2-amino-3-phenyl-1-propanol, 2r-2-amino-3-phenylpropan-1-ol, d +-phenylalaninol, +-d-phenylalaninol, r-2-amino-3-phenyl-1-propanol, d-+-phenylalaninol, r-phenylalaninol

Spécification

Melting Point 89°C to 94°C
Quantity 1 g
Optical Rotation +23 ± 2° (c=5 in Ethanol)
Sensitivity Air Sensitive
UN Number UN3259
Beilstein 4665408
Solubility Information Soluble in Dichloromethane,Ethyl Acetate,Methanol.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material D-Phenylalaninol

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis