missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D(+)-Phenyllactic acid, 98%, Thermo Scientific Chemicals
50.80 € - 168.00 €
Chemical Identifiers
CAS | 7326-19-4 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00078062 |
InChI Key | VOXXWSYKYCBWHO-MRVPVSSYSA-N |
Synonym | r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid |
PubChem CID | 643327 |
ChEBI | CHEBI:32978 |
IUPAC Name | (2R)-2-hydroxy-3-phenylpropanoic acid |
SMILES | O[C@H](CC1=CC=CC=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10796622
|
Thermo Scientific Acros
344580010 |
1 g | Glass bottle |
50.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10377751
|
Thermo Scientific Acros
344580050 |
5 g | Glass bottle |
168.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7326-19-4 | |
166.18 | |
VOXXWSYKYCBWHO-MRVPVSSYSA-N | |
643327 | |
(2R)-2-hydroxy-3-phenylpropanoic acid |
C9H10O3 | |
MFCD00078062 | |
r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid | |
CHEBI:32978 | |
O[C@H](CC1=CC=CC=C1)C(O)=O |
Specifications
7326-19-4 | |
White | |
98% | |
C6H5CH2CH(OH)COOH | |
+18.5° (20°C c=1,CH3OH) | |
r-2-hydroxy-3-phenylpropionic acid, r-2-hydroxy-3-phenylpropanoic acid, 2r-2-hydroxy-3-phenylpropanoic acid, d-+-phenyllactic acid, d-+-3-phenyllactic acid, r-phenyllactate, r-+-3-phenyllactic acid, 3-phenyl-d-lactic acid, r-3-phenyllactic acid, r-, a-hydroxy-3-phenylpropionic acid | |
O[C@H](CC1=CC=CC=C1)C(O)=O | |
166.18 | |
CHEBI:32978 | |
98% | |
D(+)-Phenyllactic acid, 98% |
121°C to 125°C | |
Authentic | |
C9H10O3 | |
MFCD00078062 | |
+ 18.5 | |
VOXXWSYKYCBWHO-MRVPVSSYSA-N | |
(2R)-2-hydroxy-3-phenylpropanoic acid | |
643327 | |
166.18 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 230-803-3
Product Identifier
- D(+)-Phenyllactic acid
RUO – Research Use Only