Learn More
Description
D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Proline is an essential component of collagen and is important for proper functioning of joints and tendons. It is also used as asymmetric catalysts in proline organocatalysis reactions.
Solubility
Soluble in water (1620 g/L at 20°C), and acetone (slightly). Insoluble in ethanol.
Notes
It is hygroscopic. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 213°C to 215°C (decomposition) |
Beilstein | 80811 |
Solubility Information | Soluble in water (1620g/L at 20°C),and acetone (slightly). Insoluble in ethanol. |
Density | 1.36 |
Quantity | 25 g |
Formula Weight | 115.13 |
Optical Rotation | +84° (c=1 in water) |
Sensitivity | Hygroscopic |
Percent Purity | 99% |
Chemical Name or Material | D-Proline |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10245314 | Certificate of Analysis | 11/01/2023 | A10682.06,A10682.03,A10682.14 |
10241625 | Certificate of Analysis | 18/07/2022 | A10682.06,A10682.03,A10682.14 |
10238029 | Certificate of Analysis | 12/04/2022 | A10682.06,A10682.03,A10682.14 |
10099566 | Certificate of Analysis | 22/11/2021 | A10682.06,A10682.03,A10682.14 |
10113828 | Certificate of Analysis | 22/11/2021 | A10682.06,A10682.03,A10682.14 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.