missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Serine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 220.0°C |
Assay Percent Range | 98% |
Linear Formula | HOCH2CH(NH2)CO2H |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: soluble in water: 346g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene., soluble in mineral acids., insoluble in organic solvents. |
Specific Rotation | − 14.75 |
Formula Weight | 105.09 |
Percent Purity | 98% |
Specific Rotation Condition | −14.75° (20°C c=10 2 N HCl) |
Quantity | 5 g |
Show More |
Safety and Handling
Product Identifier
- D-Serine
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'