missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-Sorbitol, 97%
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Spezifikation
Melting Point | 95.0°C to 99.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (on anhydrous substance) (HPLC) |
Packaging | Plastic bottle |
Quantity | 100 g |
Beilstein | 1533 |
Merck Index | 158851 |
Specific Rotation Condition | + 4.00 (20.00°C per eur. pharm.) |
Specific Rotation | + 4.00 |
Mehr anzeigen |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur