missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Sorbitol, 98%

Product Code. 11367868
Click to view available options
Quantity:
500 g
2 kg

D-sorbitol is added to food products as sweetener. It is also used in industrial and medical applications. Sorbitol often is applied in modern cosmetics as a humectant and thickener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.172
MDL Number MFCD00004708
InChI Key FBPFZTCFMRRESA-JGWLITMVSA-N
Synonym d-sorbitol, sorbitol, d-glucitol, glucitol, l-gulitol, --sorbitol, diakarmon, glucarine, sorbol, d---sorbitol
PubChem CID 5780
ChEBI CHEBI:17924
IUPAC Name (2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
SMILES C(C(C(C(C(CO)O)O)O)O)O

Specifications

Melting Point 98°C to 100°C
Assay Percent Range 98%
Quantity 2 kg
Beilstein 1721899
Sensitivity Hygroscopic
Solubility Information Soluble in water,methanol,acetone and DMF.
Formula Weight 182.17
Physical Form Powder
Chemical Name or Material D-Sorbitol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'