missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals

Product Code. 10647421
Click to view available options
Quantity:
1 kg
5 kg
25 kg

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-50-1
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006626
InChI Key CZMRCDWAGMRECN-PWPRYFECNA-N
Synonym sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
PubChem CID 5988
ChEBI CHEBI:17992
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 190.0°C to 192.0°C
Color White
Ignition Residue 0.01% max.
Loss on Drying 0.03% max. (105°C)
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 1 kg
Beilstein 31,424
Merck Index 15,9012
Specific Rotation +66.3° to +66.8° (25°C, 589nm) (c=26, H2O)
Solubility Information Solubility in water: 1970g/L (15°C). Other solubilities: moderately soluble in glycerol and pyridine
Formula Weight 342.29
Grade ACS Reagent
Insoluble Matter 0.005% max
Titratable Acid 0.0008 meq/g max
Physical Form Crystals
Chemical Name or Material D(+)-Sucrose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'