missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid |
Formula Weight | 150.09 |
Grade | BAKER ANALYZED™ ACS Reagent |
Quantity | 500 g |
Packaging | Poly Bottle |
Physical Form | Crystalline |
Specific Gravity | 1L = 1.76kg |
Chemical Name or Material | d-Tartaric Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'