missing translation for 'onlineSavingsMsg'
Learn More

d-Tartaric Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15537834
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

IUPAC Name 2,3-dihydroxybutanedioic acid
Formula Weight 150.09
Grade BAKER ANALYZED™ ACS Reagent
Quantity 2.5 kg
Packaging Poly Bottle
Physical Form Crystalline
Specific Gravity 1L = 1.76kg
Chemical Name or Material d-Tartaric Acid
compliance-icons
Product Identifier
  • L(+)-Tartaric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L(+)-Tartaric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.