missing translation for 'onlineSavingsMsg'
Learn More
D-tert-Leucine, 99%, Thermo Scientific Chemicals Product Code.: 11388965

D-tert-Leucine, 99%, Thermo Scientific Chemicals

Product Code. 11388965
2 g
Click to view available options
Quantity:
100 mg
500 mg
2 g
This item is not returnable. View return policy

Product Code. 11388965

Brand: Thermo Scientific Alfa Aesar L14098.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-tert-Leucine is used for synthesis of amino acids e.g. in the synthesis of aldehydes by double alkylation of unsaturated aldimines of tert-Leu esters.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26782-71-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00004265
InChI Key NPDBDJFLKKQMCM-UHFFFAOYNA-N
Synonym d-tert-leucine, h-d-tle-oh, d-tert-butylglycine, r-2-amino-3,3-dimethylbutanoic acid, d-t-leucine, r-tert-leucine, 2r-2-amino-3,3-dimethylbutanoic acid, d-t-butylglycine, h-tbu-d-gly-oh, d-valine, 3-methyl
PubChem CID 6950340
IUPAC Name (2R)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)C(N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721825
Solubility Information Soluble in water.
Quantity 2 g
Formula Weight 131.18
Optical Rotation −7° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-tert-Leucine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10180944 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberL14098.ME,L14098.04
Lot Number10189715 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14098.ME,L14098.04
Lot NumberK29X011 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14098.ME,L14098.04
Lot Number10160446 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14098.ME,L14098.04
Lot NumberF08X060 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14098.ME,L14098.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
D-tert-Leucine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.