missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals D-(+)-Trehalose, anhydrous
Click to view available options
Quantity:
5 g
10 g
50 g
Description
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.
Solubility
Soluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
Color | White |
Quantity | 50 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Optical Rotation | +197° (c=7 in Water) |
Formula Weight | 342.3 |
Physical Form | Powder |
Chemical Name or Material | D-(+)-Trehalose, anhydrous |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'