Learn More
Melford Daidzein
Inactive analog of Genistein.
Brand:  Melford D20010-0.025
 View more versions of this product
Product Code. 30018226
  • 67.90 € / 25mg
Explore more special offers
Description
Description
Inactive analog of Genistein.
Inactive analog of Genistein. Inhibits casein kinase II activity.
Specifications
Specifications
486-66-8
C15H10O4
25 mg