missing translation for 'onlineSavingsMsg'
Learn More

Daidzin, 98%, Thermo Scientific Chemicals

Product Code. 15569513
Click to view available options
Quantity:
100 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-66-9
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00017466
InChI Key KYQZWONCHDNPDP-QNDFHXLGSA-N
Synonym daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside
PubChem CID 107971
ChEBI CHEBI:42202
IUPAC Name 3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Loss on Drying 2.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Formula Weight 416.38
Percent Purity 98%
Chemical Name or Material Daidzin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'