missing translation for 'onlineSavingsMsg'
Learn More

DAPT, Thermo Scientific Chemicals

Product Code. 15467109
Click to view available options
Quantity:
10 mg

 

DAPT is used in the study of β-amyloid formation. DAPT has been shown to inhibit Notch signaling in studies of autoimmune and lymphoproliferative diseases, such as ALPS and lupus erythematosus, as well as in cancer cell growth, angiogenesis, and differentiation of human induced pluripotent stem cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DAPT is used in the study of β-amyloid formation. DAPT has been shown to inhibit Notch signaling in studies of autoimmune and lymphoproliferative diseases, such as ALPS and lupus erythematosus, as well as in cancer cell growth,
TRUSTED_SUSTAINABILITY

Specifications

CAS 208255-80-5
Color White
Molecular Formula C23H26F2N2O4
MDL Number MFCD04974585
Quantity 10 mg
Sensitivity Light Sensitive
Synonym dapt, dapt gsi-ix, gamma-secretase inhibitor ix, gsi-ix, insolution gamma-secretase inhibitor ix, s-tert-butyl 2-s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, n-n-3,5-difluorophenacetyl-l-alanyl-s-phenylglycine t-butyl ester, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetamido propanamido-2-phenylacetate, tert-butyl 2s-2-2s-2-2-3,5-difluorophenyl acetyl amino propanoyl amino-2-phenylacetate, gsiix
Solubility Information Soluble in DMSO,ethanol,dichloromethane,DMF.
InChI Key DWJXYEABWRJFSP-XOBRGWDASA-N
SMILES CC(C(=O)NC(C1=CC=CC=C1)C(=O)OC(C)(C)C)NC(=O)CC2=CC(=CC(=C2)F)F
IUPAC Name tert-butyl (2S)-2-[[(2S)-2-[[2-(3,5-difluorophenyl)acetyl]amino]propanoyl]amino]-2-phenylacetate
Molecular Weight (g/mol) 432.468
PubChem CID 5311272
ChEBI CHEBI:86193
Formula Weight 432.5
Physical Form Lyophilized
Chemical Name or Material DAPT
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'