Learn More
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
Brand:  Thermo Scientific™ 201145
 View more versions of this product
Product Code. 15563761
  • 2427.08 € / Each
Estimated Shipment: 07-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Integrate Thermo Scientific™ Data Output Accessories for your environmental chamber or incubator to increase functionality and compatibility.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single Pen, 6 in. 7 Day Chart Recorders; 0° to 60°C (32° to 140°F)
LED
Accessory, Chart Recorders and Paper
Environmental Chamber (3948 and 3907)
0°C to 60°C
Customer Installed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Data Output Accessories for Environmental Chambers and Incubators >