Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Brand:  Thermo Scientific™ 195897
 View more versions of this product
Product Code. 11796649
  • 563.81 € / Each
Estimated Shipment: 01-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RS-485; 4-20mA
Revco™ CxF, HERAfreeze™ HFC, Model 1535 Monitor/Alarm System, TSC Series
4 to 20 mA
Output
Alarm, Recorder