Learn More
Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Brand:  Thermo Scientific™ 195895
 View more versions of this product
Product Code. 11973937
  • 448.23 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RS-232; 0-1V
Forma™ 8600, HERAfreeze™ HFC, Revco™ CxF, TSC Series
0/1 V
Refrigerator/Freezer Electronics
Alarm, Recorder