Learn More
Cytiva Datrys Life Science Software for Biochrom Spectrophotometers
Datrys Life Science software is used for nucleic acid, protein, and cell density measurements and analysis. Compatible with Biochrom Ultrospec 7000/8000/9000 UV-Vis Spectrophotometers.
Brand:  Cytiva 29003623
Product Code. 12959820
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Wavelength scanning, kinetics, quantitative calibration curves
  • Life science methods
  • Validation
  • Method Developer
  • Accessory control
Specifications
Specifications
Datrys Life Science Software
For nucleic acids, proteins, and cell density measurements
Spectrophotometers