missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Dave S3 Shoe

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Dave.

133.00 €

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Brown
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
View More Specs

Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17582616
View Documents
Emma Safety Footwear
MM96654939
39
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17592616
View Documents
Emma Safety Footwear
MM96654940
40
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17502626
View Documents
Emma Safety Footwear
MM96654941
41
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17512626
View Documents
Emma Safety Footwear
MM96654942
42
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17522626
View Documents
Emma Safety Footwear
MM96654943
43
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17532626
View Documents
Emma Safety Footwear
MM96654944
44
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17542626
View Documents
Emma Safety Footwear
MM96654945
45
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17552626
View Documents
Emma Safety Footwear
MM96654946
46
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17562626
View Documents
Emma Safety Footwear
MM96654947
47
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
17572626
View Documents
Emma Safety Footwear
MM96654948
48
133.00 €
1 pair
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
Description

Description

  • S3-Safetyshoe with sole plate for wet work
  • Meral Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Dave.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications

Specifications

ISO 20345:2011
Brown
Hydro-Tec Antibacterial
Standard Width
Safety Shoe
Laces
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Nubuck, Oiled Nubuck
Low-cut
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.