Learn More
Tocris Bioscience™ DBHDA
Reagent for synthetic biology; converts cysteine to dehydroalanine
Brand:  Tocris Bioscience™ 6175/50
Product Code. 18776628
  • 73.00 € / 50mg
Explore more special offers
Description
Description
DBHDA is a reagent for synthetic biology; converts cysteine to dehydroalanine (Dha).
Specifications
Specifications
Synthetic Biology Reagent
C6H10Br2N2O2
Store at 4°C
50 mg
99584-96-0
301.96g/mol
Buffers, Chelators and Reagents