Learn More
VACUUBRAND™ DCP 3000 + MPT Vacuum Gauge Set
Monitor up to eight external vacuum sensors with this vacuum gauge system. Vacuubrand™ DCP 3000 + MPT Vacuum Gauge Set includes a vacuum gauge and an external vacuum sensor MPT.
Brand: VACUUBRAND™ 20683175
This item is not returnable.
View return policy
Description
- Ready for operation, with wall-plug power supply and manual.
- For the range from atmospheric pressure down to high vacuum (5 x 10-9mbar)
- Kit includes combined thermal conductivity / cold cathode (Pirani / Penning) vacuum sensor MPT.
- Can be used for relative pressure measurements (using VSK 3000 vacuum sensors as a reference) and as a data logger for up to 32,000 measurement values.
- Uniform plug and socket connections make the system flexible and allow extension of the lines up to 30m.
- Components connected to the gauges and controllers are automatically recognized, configured and supervised.
- For high vacuum measurements, the DCP 3000 can be connected with the combined Penning/Pirani gauge head MPT.
Specifications
100-230V, 50-60Hz | |
114 mm | |
144 mm | |
0.44 kg | |
Vacuum Gauge Set |
144 x 124 x 114 mm | |
Stainless steel tubing KF DN 25, Vent valve VBM-B, Vacuum sensor VSK 3000, Vacuum sensor VSP 3000, Vacuum sensor MPT 200 | |
10°C to 40°C | |
1 x 10–8 - 2 x 10–3 mbar: ±25%; 2 x 10–3 - |
Product Content Correction
Product Title
VACUUBRAND™ DCP 3000 + MPT Vacuum Gauge Set > Dimensions (L x W x H): 144 x 124 x 114mm
Spot an opportunity for improvement?Share a Content Correction