missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCTPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | DCTPP1 |
---|---|
Concentration | 0.38 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836394
|
Proteintech
16684-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826394
|
Proteintech
16684-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is dCTP pyrophosphatase, which converts dCTP to dCMP and inorganic pyrophosphate. The encoded protein also displays weak activity against dTTP and dATP, but none against dGTP. This protein may be responsible for eliminating excess dCTP after DNA synthesis and may prevent overmethylation of CpG islands. Three transcript variants, one protein-coding and the other two non-protein coding, have been found for this gene.Specifications
DCTPP1 | |
Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
Q91VC0, Q9H773, Q9QY93 | |
192252, 66422, 79077 | |
DCTPP1 Fusion Protein Ag10015 | |
Primary | |
-20°C | |
DCTPP1 |
0.38 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDA03, dCTP pyrophosphatase 1, dCTPase 1, DCTPP1, Deoxycytidine triphosphatase 1, RS21C6, XTP3TPA | |
Dctpp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |