Learn More
Thermo Scientific™ Deactivated Glass Wool
Pack GC injection port liners with deactivated glass wool for good homogenization, volatilization of high boiling point compounds, and minimal column contamination.
75.60 €
Specifications
Packing Material Deactivated Glass Wool
For Use With (Equipment) GC Liners
View More Specs
Products 1
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11567781
View Documents
Thermo Scientific™
60180-785
Deactivated Glass Wool 10 g
75.60 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
Description
Description
Pack your GC injection port liner with Thermo Scientific™ deactivated glass wool for good homogenization and volatilization of high boiling point compounds within the liner. The glass wool acts as a barrier to prevent non-volatile sample matrix compounds and septa particles from entering GC columns, minimizing contamination and increasing precision and reproducibility.
Specifications
Specifications
Deactivated Glass Wool
GC Liners