missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11324529
Click to view available options
Quantity:
10,000 mL
100 mL
2500 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

Solubility
Miscible with acetone, benzene, ether, methanol, aniline, decalin, chloroform, butanol and ethanol. Immiscible with water.

Notes
Store in cool place. Air, light and heat sensitive. Heat and light accelerate peroxide formation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point <-45°C
Density 0.88
Boiling Point 189°C to 191°C
Flash Point 57°C (134°F)
Odor Mild
Refractive Index 1.474
Quantity 100 mL
UN Number UN1147
Beilstein 878165
Merck Index 14,2846
Solubility Information Miscible with acetone,benzene,ether,methanol,aniline,decalin,chloroform,butanol and ethanol. Immiscible with water.
Formula Weight 138.25
Percent Purity 98%
Chemical Name or Material Decahydronaphthalene, cis + trans
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'