missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene, cis + trans, 98%, Thermo Scientific Chemicals

Product Code. 11441578
500 mL
Click to view available options
Quantity:
10,000 mL
100 mL
2500 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decahydronaphthalene is widely used as an industrial solvent for resins and fuel additives. It is a substitute for turpentine in lacquers, shoe polishes and waxes.

Solubility
Miscible with acetone, benzene, ether, methanol, aniline, decalin, chloroform, butanol and ethanol. Immiscible with water.

Notes
Store in cool place. Air, light and heat sensitive. Heat and light accelerate peroxide formation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-17-8
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00004130
InChI Key NNBZCPXTIHJBJL-UHFFFAOYSA-N
Synonym decalin, decahydronaphthalene, cis-decahydronaphthalene, trans-decahydronaphthalene, cis-decalin, dekalin, naphthalene, decahydro, perhydronaphthalene, trans-decalin, naphthalane
PubChem CID 7044
ChEBI CHEBI:38853
IUPAC Name 1,2,3,4,4a,5,6,7,8,8a-decahydronaphthalene
SMILES C1CCC2CCCCC2C1

Specifications

Melting Point <-45°C
Density 0.88
Boiling Point 189°C to 191°C
Flash Point 57°C (134°F)
Odor Mild
Refractive Index 1.474
Quantity 500 mL
UN Number UN1147
Beilstein 878165
Merck Index 14,2846
Solubility Information Miscible with acetone,benzene,ether,methanol,aniline,decalin,chloroform,butanol and ethanol. Immiscible with water.
Formula Weight 138.25
Percent Purity 98%
Chemical Name or Material Decahydronaphthalene, cis + trans
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Decahydronaphthalene, cis + trans, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.