missing translation for 'onlineSavingsMsg'
Learn More

Decahydronaphthalene-d18, for NMR, packaged in ampoules, 98+% atom D, Thermo Scientific™

Product Code. 10003590
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

For NMR
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Color Colorless
Deuterium (D) 98.0% min.
Quantity 1 g
CAS 28788-42-3
Molecular Formula C10H18
Linear Formula C10D18
MDL Number MFCD00044371
Synonym cis-decahydronaphthalene-d18, decahydronaphthalene-d18, decalin™-d18, d18-cis-decahydronaphthalene, decahydro-d10-naphthalene-d8, decahydronaphthalene-d18 cis/trans, decahydronaphthalene-d18, 98 atom % d, cis-decahydronaphthalene-d18,99.5 atom % d cis trans
InChI Key NNBZCPXTIHJBJL-XRRAJICISA-N
SMILES C1CCC2CCCCC2C1
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecadeuterionaphthalene
Molecular Weight (g/mol) 156.364
PubChem CID 2724146
Formula Weight 156.4
Grade NMR
Chemical Name or Material Decahydronaphthalene-d18, 98+ atom% D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Decahydronaphthalene-d18, packaged in ampules
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Decahydronaphthalene-d18, for NMR, packaged in ampoules, 98+% atom D, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.