missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decanoic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
Melting Point | 30.0°C to 32.0°C |
Boiling Point | 268.0°C to 270.0°C |
Color | White |
Assay Percent Range | 99% |
Linear Formula | CH3(CH2)8CO2H |
Beilstein | 02, II, 309 |
Merck Index | 15, 1760 |
Solubility Information | Solubility in water: 0.15g/L (20°C). Other solubilities: soluble in dilute hno3, soluble in ethanol, ether, chloroform, benzene and, carbon disulfide |
Formula Weight | 172.27 |
Percent Purity | 99% |
Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Decanoic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P362-Take off contaminated clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'