Learn More
Fisherbrand™ Decapping Heads for Programmable Electronic High Power Crimp Station
For use with Programmable Electronic High Power Crimp Station
368.00 € - 414.12 €
Specifications
Material Plastic, Steel
Autoclavable Not Autoclavable
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15552310
View Documents
Fisherbrand™
11 07 3203
Decapping Head
368.00 €
Each
Estimated Shipment: 04-12-2024
to see stock.
Add to basket
15542340
View Documents
Fisherbrand™
20 07 3207
20mm Decapping Head This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Decapping Head for Alumnium/Steel Caps
  • Not adjustable
Specifications
Specifications
Plastic, Steel
Not Autoclavable