Learn More
Decitabine, MedChemExpress
MedChemExpress Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.
POA - 66.15 €
Specifications
Chemical Name or Material | Decitabine |
---|---|
CAS | 2353-33-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C8H12N4O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16487976
|
Medchem Express
HY-A0004 |
100 mg |
66.15 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16428258
|
Medchem Express
HY-A0004 |
10 mg |
POA
10mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16404147
|
Medchem Express
HY-A0004 |
50 mg |
POA
50mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Decitabine (NSC 127716) is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. Decitabine incorporates into DNA in place of cytosine can covalently trap DNA methyltransferase to DNA causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis. Decitabine has potent anticancer activity.Specifications
Decitabine | |
White | |
C8H12N4O4 | |
DMSO : ≥ 50 mg/mL (219.10 mM) ∣H2O : 20 mg/mL (87.64 mM; Need ultrasonic) | |
228.21 | |
95.0% | |
Solid |
2353-33-5 | |
Research | |
5-Aza-2'-deoxycytidine 5-AZA-CdR NSC 127716 | |
O=C1N=C(N)N=CN1[C@H](O[C@@H]2CO)C[C@@H]2O | |
228.21 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only