missing translation for 'onlineSavingsMsg'
Learn More

Decon™ Decomatic

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines

221.00 €

Specifications

Form Liquid
pH >13
Appearance Amber
Viscosity 2.28cps at 20°C
View More Specs

Products 1
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10538610
View Documents Promotion Details
Decon™
DEC10
10 L
221.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines. By preventing the deposition of lime and salts on both contents and machine parts, it minimises corrosion and wear of seals, pumps, racks and stainless steel surfaces, thereby providing more efficient performance and longer life to the machine. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Ideal for use on glassware and equipment in medical, pharmaceutical, research, educational, scientific and industrial laboratories.
Specifications

Specifications

Liquid
>13
Amber
2.28cps at 20°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DECON DECOMATIC

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains : NaOH

SDS
Documents

Documents

Product Certifications