missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Deep-Form Photographic Tray without Bottom Ribs

Use these trays for universal laboratory or storage purposes. Buerkle™ Deep-Form Photographic Tray without Bottom Ribs are made of high quality PVC and available in shallow or deep models in 3 attractive, high-gloss colors: white, red and black.

Brand:  Buerkle™ 4207-2040

104.92 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10580312

  • 123.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • PVC
  • Thick sides, shatterproof
  • Largely resistant to acids and alkalis
  • Liquid can be poured out easily from the tray edges
  • Not suitable for temperature controlled cupboards or dishwashers
  • Temperature resistant from -30 to +60°C
Specifications

Specifications

Red
420 mm (Inner Base), 520 mm (Outer Base)
Photographic Tray
4.33 in.
110 mm
PVC
510 mm (Inner Base), 610 mm (Outer Base)
Rectangular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers