missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Non-sterile Deep Well Plate for Storage and Collection

Deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.

Brand:  Corning™ 3866

 View more versions of this product

Product Code. 16862250

  • 569.00 € / Pack of 50
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of polypropylene
  • Offer excellent chemical resistance
  • Can withstand temperatures down to -80°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Without Lid
Polypropylene
Non-sterile
2 mL
Translucent
Clear
96
Non-treated
Round
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers